missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 10,000 Class 2, Level A Suits

Catalog No. 191300004
Change view
Click to view available options
Size:
Large
X-Large
Style:
Front Entry
Rear Entry
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
191300004 Large Rear Entry
191300002 X-Large Front Entry
191300001 Large Front Entry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 191300004 Supplier DuPont™ Supplier No. TK613TLYLG000100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extremely durable to abrasion, tearing, punctures, and cuts

Tychem TK Class 2 Level A Suits—Use NFPA 1994 Class 2 suits in situations where an unidentified chemical/biological substance of unknown concentration poses an immediate threat.

  • Extra-wide three-layer faceshield
  • Double taped seams
  • Gastight zipper closure
  • Double storm flaps
  • Attached three-layer gloves and sock boots
  • High-visibility lime yellow
Tychem Responder NFPA 1994 Class 2 Suits—offer a broad range of chemical protection. Patented dark blue fabric consists of multiple film barriers with unique layers laminated to each side of a polypropylene substrate.
  • Gastight zipper lock closure with double storm flaps
  • Double-taped seams
  • Two-layer 40mil PVC faceshield
  • Three exhaust valves
  • Attached three-layer glove system
  • Attached sock boots with outer boot flaps

Compliance

Meets NFPA 1994 Standard on Protective Ensembles for First Responders to Hazardous Materials Emergencies and CBRN Terorrism Incident, Class 2 requirements.

NOTE: Product safety information supplied corresponds to current knowledge on the subject. This information may be subject to revisions as new knowledge and experience becomes available. DuPont makes no warranties and assumes no liability in connection with any use of this information. It is the end user's responsibility to determine level of risk and select proper protective equipment for end-use conditions. Nothing in this product offering is to be considered license to operate under or a recommendation to infringe upon any patent rights.

Specifications

Certifications/Compliance NFPA 1994, Class 2
Color Lime Yellow
For Use With (Application) Industrial, HazMat and domestic preparedness applications
Material Tychem
Size Large
Closure Type Gas-tight zipper
Description Expanded faceshield, 2 exhaust valves, internal adjustment waist belt, attached internal multilayer laminate/external Neoprene size 10 gloves, socks with outer boot flaps, soles; Specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals, excellent chemical barrier properties and offers an extremely durable Fabric that is puncture and tear-resistant
Height (English) 5.9 to 6.3 ft.
Height (Metric) 1.79 to 1.92 m
Length (English) Inseam 31 in.
Length (English) Sleeve 29 in.
Length (Metric) Inseam 78.74 cm
Length (Metric) Sleeve 73.66 cm
Seam Type Taped
Size (English) Chest 46.75 to 50.25 in.
Size (Metric) Chest 118.75 to 127.65 cm
Style Rear Entry
Thickness 27 mil
Show More Show Less