Learn More
Coast TX17R Tactical Rechargeable Plus Flashlight
Catalog No. 17010308
Encompass
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
17010308 LED
1 options
Catalog No. 17010308 Supplier Coast Supplier No. 30778
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The rechargeable TX17R, ideal for dynamic beam control in the medium-to-long range, flexes the full capabilities of the COAST PURE BEAM™ system-delivering seamless transitions between Ultra View Flood Beam and BULLS-EYE™ Spot Beam.
  • Maxing out at 1250 lumens with a 234-meter (767-foot) beam, this three-mode flashlight (medium, high, low) is just under 6 inches long and fitted with a reversible pocket clip to make it the ultimate carry-everywhere, do-anything lighting solution
  • Control the beam shape with subtle pushes and pulls using the SLIDE FOCUS™ system, or simply twist the bezel to set the beam where you like it with the BEAM LOCK™
  • To support hands-free operation, the anti-roll bezel prevents the flashlight from moving when placed on uneven surfaces
  • From full power, the TX17R can run up to 21 hours on low and then recharges via USB-C
  • The COAST Rechargeable Plus system allows the flashlight to run on its ZITHION-X™ ZX850 lithium battery or on two CR123 disposable lithium batteries (purchased separately) for added convenience
  • Machined from solid aluminum, the TX17R is built to IP54 specifications for water resistance and dust resistance and is backed by the COAST lifetime warranty
Specifications
Light Source LED
Voltage 3.7 V
Product Type LED Flashlight