missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TX-750 Swinging Bucket/BIOShield 1000A Adapters

Catalog No. 75003737
Change view
Click to view available options
Capacity:
1.5/2.0 mL Conical Microtube
10 mL Blood Collection/15mL DIN Tube
100 mL Round-bottom Open-top Tube
14 mL Flanged Round-bottom Tube
25 mL DIN Round-bottom Tube
25 mL Universal Tube
250 mL Flat-bottom Bottles
5/7 mL Blood Collection Tube
5/7 mL Round-bottom Tube
50 mL Round-bottom Tube
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75003737 250 mL Flat-bottom Bottles
75003742 100 mL Round-bottom Open-top Tube
75003749 50 mL Round-bottom Tube
75003755 25 mL Universal Tube
75003756 25 mL DIN Round-bottom Tube
75003758 14 mL Flanged Round-bottom Tube
75003767 10 mL Blood Collection/15mL DIN Tube
75003768 5/7 mL Blood Collection Tube
75003769 5/7 mL Round-bottom Tube
75003770 1.5/2.0 mL Conical Microtube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 75003737 Supplier Thermo Scientific™ Supplier No. 75003737

Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.

Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.

  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
  • Maximum tube size: 63 × 130mm

Specifications

Type Swinging Bucket Adapter
Capacity 250 mL Flat-bottom Bottles