Learn More
Thermo Scientific™ TX-750 Rectangular Bucket Adapters
Catalog No. 75003685
Change view
Click to view available options
Capacity:
15 mL Conical Tubes
50 mL Conical Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75003685 50 mL Conical Tubes
75003684 15 mL Conical Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75003685 Supplier Thermo Scientific™ Supplier No. 75003685
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Ordering Information:
Order rectangular buckets (75003614), Thermo Scientific™ ClickSeal™ Biocontainment Lids (75003615) and TX-750 Rotor (75003607) separately. Contact your sales representative for more information.
Specifications
Type Tube Adapter
Capacity 50 mL Conical Tubes