Learn More
Thermo Scientific™ TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Supplier: Thermo Scientific™ 75003802
Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.- Maximum tube capacity: 4 × 100mL
- Maximum O.D. × L: 48 × 122mm
Chemical Identifiers
| Rotor Tube Adapter |
Specifications
| Rotor Tube Adapter | |
| 50 mL DIN Round-bottom Tube | |
| 35 mm | |
| 126 mm |