Learn More
Thermo Scientific™ TX-200 Rotor Tube Adapters
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Supplier:  Thermo Scientific™ 75003785
 View more versions of this product
Catalog No. 75003785
Only null left
Description
Description
Use Thermo Scientific™ TX-200 Rotor Tube Adapters with round buckets in the TX-200 Rotor.
Adapters hold a variety of tubes including round-bottom, conical, urine, blood collection and microtubes.
  • Maximum tube capacity: 4 × 100mL
  • Maximum O.D. × L: 48 × 122mm
Specifications
Chemical Identifiers
Rotor Tube Adapter
Specifications
Rotor Tube Adapter
5/7 mL Blood Collection Tube
13 mm
122 mm
Product Suggestions
Product Suggestions