Learn More
Thermo Scientific™ ClickSeal Biocontainment Lids for TX-1000 Rotor, set of 4
Catalog No. 75007309
Change view
Click to view available options
TX-1000 ClickSeal Biocontainment Lids (set of 4)
Catalog No.
75007309 TX-1000 ClickSeal Biocontainment Lids (set of 4)
1 options
Catalog No. 75007309 Supplier Thermo Scientific™ Supplier No. 75007309
Process 4L in a traditional 3L general purpose centrifuge footprint with the TX-1000 rotor.
Process 4L in a traditional 3L general purpose centrifuge footprint with the Thermo Scientific™ TX-1000 rotor. High-throughput capacities for processing more tubes in less runs saves time while reducing energy and wear. The rotor features Thermo Scientific™ Slide Coat™ finish, an advanced lubricant and corrosion resistant coating. Dual-purpose large bucket design accepts adapters for tubes or microplates. Save time: No need to swap buckets. Save space: No additional buckets to store. Save money: No additional buckets to purchase.
Key features include:
  • Spin up to 40 x 50 mL conical tubes, 196 blood tubes or 24 microplates
  • High capacities process more tubes in less runs, saving time while reducing energy and wear
  • Slide Coat finish offers an advanced lubricant and corrosion resistant coating
  • Dual-purpose large bucket design accepts adapters for tubes or microplates
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
Ordering Information:
Buckets, lids and microplate carriers are sold separately; order Rotor Body (75-003-607) and components to complete assembly. For Replacement O-Rings for lids (Set of 4) order 75-003-610. Tube Adapters are also available separately. Contact your Sales Representative for information.
Specifications
For Use With (Equipment) Thermo Scientific Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40 and SL 40/40F Centrifuges
Description ClickSeal Biocontainment Lids for TX-1000 Rotor (set of 4)
Certifications/Compliance Biocontainment certification by Public Health England, Porton Down, U.K.
Product Type Bucket Lid