Learn More
Selleck Chemical Llc TWS119 10mg 601514-19-6
Supplier: Selleck Chemicals Co Ltd S159010MG
TWS119 is a GSK-3? inhibitor with IC50 of 30 nM in a cell-free assay; capable of inducing neuronal differentiation and may be useful to stem cell biology. GSK-3? inhibition triggers autophagy. *For Research & Development use only. Product is not intended for drug, household, or human consumption.