Learn More
United Scientific™ Two-Way Burner Stand
Catalog No. S15703
Change view
Click to view available options
For Use With (Equipment):
Alcohol Burners
Catalog No. For Use With (Equipment)
S15703 Alcohol Burners
1 options
Catalog No. S15703 Supplier United Scientific™ Supplier No. ABHL01
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Unique design allows the stand to be used in high or low positions.
  • This stand is designed for use with alcohol burners.
  • Designed for use with alcohol burners
  • Base constructed from 3/16 in. stainless steel rod, with plated steel mesh top
  • Can be positioned to height of either 4 or 5 in.
Specifications
Product Type Stand
For Use With (Equipment) Alcohol Burners
Material Steel