Learn More
Eisco™ Two-Stroke Diesel Engine Model
Catalog No. S23748
Change view
Click to view available options
9L x 9W x 1.5 in. H
Catalog No.
S23748 9L x 9W x 1.5 in. H
1 options
Catalog No. S23748 Supplier Eisco™ Supplier No. PH0490
Demonstrates operating principles of an air cooled two-stroke engine.
  • Shows internal structure
  • Aluminum and plastic parts
  • Ignition shown via small bulb
  • Sectioned carburetter and fuel supply
  • Hand crank provided for manual operation
  • Mounted on base
Specifications
Product Type Model, Two-Stroke Diesel Engine
Description Shows internal structure
For Use With (Application) Demonstrates operating principles of an air cooled two-stroke engine
Dimensions (L x W x H) 9 x 9 x 1.5 in.