missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Two-Hole Twistit™ Rubber Stoppers
Description
- Simple twist-off seals provide convenient one, two, or three-hole use
- Solid stoppers are vacuum-tested at 30 in. of mercury
- High resistance to deformation—even after repeated autoclaving
- Each stopper has its size number in raised figures on top
Specifications
Specifications
| Product Type | Twistit Stopper |
| Stopper Number | 3 |
| Size | 3 |
| Total Positions | 2 |
| Material | Natural Rubber |
| Color | White |
| For Use With (Application) | Multi-purpose applications |