missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ Trace PCR Plates - PCR Plates

Catalog No. 01670719
Change view
Click to view available options
Color:
Colorless
Crystal Blue
Fuchsia
Product Type:
Trace PCR Plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type
01670719 Colorless Trace PCR Plate
01670718 Crystal Blue Trace PCR Plate
01670720 Fuchsia Trace PCR Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01670719 Supplier Eppendorf™ Supplier No. 0030129768

The Eppendorf twin.tec™ Trace PCR Plates feature an OptiTrack™ matrix and optical guiding grid for effortless well orientation and laser-engraved lot numbers for enhanced traceability.

  • Laser-engraved lot number and expiration date on each single plate to improve traceability in your laboratory processes
  • Unique laser-engraved optical guiding grid
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
  • Exceptionally solid, torque- and heat-resistant polycarbonate frame: No warping, bending or breaking during heat incubation or centrifugation
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Specifications

Barcode No Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 5 Plates
Product Line Eppendorf
Color Colorless
Product Type Trace PCR Plate
Skirt Style Skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 25 Pk.
Well Color Colorless
Fit for Purpose PCR Ready
Show More Show Less