missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
Supplier: Eppendorf™ 0030129636
Description
- White wells for better reflection.
- High mechanical stability.
- Effective sealing by raised well rims.
- Skirted and semi-skirted frames.
- Optimal heat transfer due to reduced wall thickness.
- Autoclavable (121°C, 20 min.).
- Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
Specifications
| Standard | |
| Autoclavable | |
| White | |
| Skirted | |
| 150 μL | |
| Non-sterile | |
| 6000 x g | |
| 10 Pk. | |
| White |
| 8 x 12 | |
| 10 x Individually Blister Wrapped | |
| PCR Plate | |
| Polypropylene, Polycarbonate | |
| 96 | |
| Real-time PCR | |
| Forensic DNA Grade (Non-sterile) | |
| Yes |