Learn More
Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates
Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.
Supplier:  Eppendorf™ 0030132718
 View more versions of this product
Catalog No. 361023840
Only null left
Description
Description
  • White wells for better reflection.
  • High mechanical stability.
  • Effective sealing by raised well rims.
  • Skirted and semi-skirted frames.
  • Optimal heat transfer due to reduced wall thickness.
  • Autoclavable (121°C, 20 min.).
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.
Specifications
Chemical Identifiers
Standard
Autoclavable
96
6000 x g
White
8 x 12
PCR Plate
Non-sterile
Yes
Specifications
Includes Barcode
8 x 12
5 Bags x 4 Plates
PCR Plate
Polypropylene, Polycarbonate
96
RT-PCR applications
PCR Clean (Non-sterile)
Yes
Standard
Autoclavable
Blue
Unskirted
150 μL
Non-sterile
6000 x g
20 Pk.
White
Product Suggestions
Product Suggestions