missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ twin.tec™ real-time qPCR Plates, 96-well - qPCR Plates

Catalog No. 361023839
Change view
Click to view available options
Color:
Blue
Blue, White Bottom
White
Skirt Style:
Semi-skirted
Skirted
Unskirted
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
Volume (Metric) Well:
150 μL
250 μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style Purity or Quality Grade Volume (Metric) Well
361023839 White Unskirted PCR Clean (Non-sterile) 150 μL
361023840 Blue Unskirted PCR Clean (Non-sterile) 150 μL
E951022015 White Skirted PCR Clean (Non-sterile) 150 μL
E951022003 Blue Skirted PCR Clean (Non-sterile) 150 μL
E951022055 White Semi-skirted PCR Clean (Non-sterile) 250 μL
E951022043 Blue, White Bottom Semi-skirted PCR Clean (Non-sterile) 250 μL
E0030129636 White Skirted Forensic DNA Grade (Non-sterile) 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 361023839 Supplier Eppendorf™ Supplier No. 0030132700

Eppendorf twin.tec™ real-time PCR plates, designed for low-volume qPCR, utilize white wells to enhance fluorescent light reflection, improving sensitivity and reproducibility in assays for reliable results.

  • White wells for better reflection.
  • High mechanical stability.
  • Effective sealing by raised well rims.
  • Skirted and semi-skirted frames.
  • Optimal heat transfer due to reduced wall thickness.
  • Autoclavable (121°C, 20 min.).
  • Available in lot-certified PCR clean and Eppendorf Forensic DNA Grade for tested purity.

Specifications

Barcode Includes Barcode
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging 5 Bags x 4 Plates
Color White
Product Type PCR Plate
Skirt Style Unskirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 150 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) RT-PCR applications
Max. RCF 6000 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 20 Pk.
Rimmed Yes
Well Color White
Show More Show Less