missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 60 (=Polyoxyethylene Sorbitan Monostearate), TCI America™
Supplier: TCI America T0545500G
Specifications
| Tween 60 (=Polyoxyethylene Sorbitan Monostearate) | |
| 9005-67-8 | |
| 500 g | |
| MFCD01780110 | |
| DEKFZWMENCCOFU-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| Yellow | |
| Crystal-Lump at 20°C | |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
| Polyoxyethylene Sorbitan Monostearate | |
| CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO | |
| 522.68 |
Chemical Identifiers
| 9005-67-8 | |
| 522.68 | |
| DEKFZWMENCCOFU-UHFFFAOYNA-N | |
| 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate |
| (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6 | |
| MFCD01780110 | |
| Polyoxyethylene Sorbitan Monostearate | |
| CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO |
Safety and Handling
EINECSNumber : (8)-0055
RTECSNumber : WG2934000
TSCA : Yes