missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Fisher BioReagents™

Catalog No. p-29380
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP337100 100 mL Glass Bottle
BP337500 500 mL HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP337100 Supplier Fisher BioReagents Supplier No. BP337 100
Only null left

Non-ionic detergent | CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol

Non-ionic detergent

This nonionic detergent is used in the isolation of water-soluble membrane proteins.

Specifications

Chemical Name or Material Polysorbate 20
Color Amber
CAS 9005-64-5
Physical Form Viscous Liquid
Boiling Point 100°C
Density 1.100g/cm³
pH 6
Quantity 100 mL
Hydroxyl Value 96 to 108
Identification Pass Test
Ignition Residue 0.25% max.
Packaging Glass Bottle
Viscosity 400 mPa/s at 25°C
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Water 3% max.
MDL Number MFCD00165986
Synonym Polyoxyethylene-20-sorbitan Monolaurate, Polyoxyethylenesorbitan monolaurate
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Molecular Weight (g/mol) 522.68
PubChem CID 443314
Show More Show Less