missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tween 20, 0.05% Aqueous, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Ricca Chemical Tween 20 0.05% (W/V) |
| CAS | 7732-18-5, 9005-64-5 |
| CAS Min % | 98.0, 0.05 |
| CAS Max % | 100.0, 0.06 |
| Concentration | 0.05% w/v |
| Quantity | 500 mL |
| Packaging | Amber Glass Bottle |
| Grade | Laboratory |