Learn More
TWD Scientific™ Tradewinds Direct Weighing Funnels
Disposable polypropylene funnels in four different sizes
$1,775.82 - $4,161.68
Specifications
| Material | Polypropylene |
|---|---|
| For Use With (Application) | Serve as weighing dishes plus sample transfer funnels |
| Product Type | Tradewinds Direct Weighing Funnel |
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
17988491
|
TWD Scientific™
DPWFPP1S |
Size: Small |
Case of 1100 Each for $4,161.68
|
|
|||||
17988492
|
TWD Scientific™
DPWFPP2M |
Size: Medium |
Case of 600 Each for $3,326.29
|
|
|||||
17988493
|
TWD Scientific™
DPWFPP3L |
Size: Large |
Case of 160 Each for $2,006.39
|
|
|||||
17988494
|
TWD Scientific™
DPWFPP4XL |
Size: X-Large |
Case of 60 Each for $1,775.82
|
|
|||||
Description
Disposable polypropylene funnels in four different sizes
- Designed to lie flat on a weighing balance Place funnel directly on balance, weigh out needed solids or liquids, transfer sample into flask
- Pouring necks designed to easily fit in volumetric flasks and test tubes
Order Info
Small funnels fit 5, 10, 25 and 50mL volumetric flasks. Medium funnels fit 100, 200 and 250mL flasks. Large funnels fit 500mL and 1L flasks. X-Large funnels fit 1 and 2L flasks.
Specifications
| Polypropylene | |
| Serve as weighing dishes plus sample transfer funnels | |
| Tradewinds Direct Weighing Funnel |