Learn More
Impacto™ TURBOTOE™ Steel Safety Toe Cap
Impacto™ TURBOTOE Steel Safety Toe Cap protects the toe area from impact, crushing or stubbing in construction and heavy work environments that carries a risk of toe damage
Supplier: Impacto™ TTUXL
Description
Impacto™ Turbotoe steel toe cap overshoes
- Made of flexible waterproof that do not harden or crack
- Treads under sole help prevent slipping
- For visitors, casual or temporary workers
- Fits tightly over existing shoes, providing a solid protecting cap
- Comfortably worn over low heel, dress, running shoes and most boots
- Color-coded for sizing
- Meet ASTM F2412/F2413 impact and compression standard
Specifications
| CE | |
| PVC | |
| Steel | |
| Yes |
| Blue | |
| X to Large, Men: 12 to 13, Women: 13 to 14 | |
| Unisex | |
| Turbotoe Safety Toe Cap |