Learn More
Impacto™ TURBOTOE™ Steel Safety Toe Cap
Catalog No. 19400029
Change view
Click to view available options
Size:
2X to Large, Men: 13 to 14
2X to Small, Men: 2 to 3, Women: 4 to 5
Large, Men: 10 to 11, Women: 12 to 13
Medium, Men: 8 to 9, Women: 10 to 11
Small; Men: 6 to 7, Women: 8 to 9
X to Large, Men: 12 to 13, Women: 13 to 14
Color:
Blue
Green
Purple
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19400029 2X to Small, Men: 2 to 3, Women: 4 to 5 Purple
19400031 Small; Men: 6 to 7, Women: 8 to 9 White
19400032 Medium, Men: 8 to 9, Women: 10 to 11 Yellow
19400033 Large, Men: 10 to 11, Women: 12 to 13 Red
19400034 X to Large, Men: 12 to 13, Women: 13 to 14 Blue
19400035 2X to Large, Men: 13 to 14 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19400029 Supplier Impacto™ Supplier No. TTUXXS
Impacto™ TURBOTOE Steel Safety Toe Cap protects the toe area from impact, crushing or stubbing in construction and heavy work environments that carries a risk of toe damage
Impacto™ Turbotoe steel toe cap overshoes
  • Made of flexible waterproof that do not harden or crack
  • Treads under sole help prevent slipping
  • For visitors, casual or temporary workers
  • Fits tightly over existing shoes, providing a solid protecting cap
  • Comfortably worn over low heel, dress, running shoes and most boots
  • Color-coded for sizing
  • Meet ASTM F2412/F2413 impact and compression standard
Specifications
Certifications/Compliance CE
Color Purple
Material PVC
Size 2X to Small, Men: 2 to 3, Women: 4 to 5
Toe Material Steel
Gender Unisex
Waterproof Yes
Product Type Turbotoe Safety Toe Cap