missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ TurboCells™ Competent Cells

Catalog No. C301020
Encompass
Change view
Click to view available options
Cell Type:
E. coli
F' E. coli
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
C301020 F' E. coli
C300020 E. coli
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C301020 Supplier Genlantis™ Supplier No. C301020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

E. coli transformation in three minutes

  • High transformation efficiencies
  • No hour-long recovery step and no inconvenient 42°C water bath
  • Useful genotypes
  • Convenient and efficiency-preserving single-use packaging

Specifications

Cell Type F' E. coli
For Use With (Application) Transformation
Includes TurboCells F genotype, Transformation Buffer, pUC19 positive control plasmid
Quantity 20 x 50 μL