missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Turbidity Standard, 1 NTU/NTRU, 60 mL

Catalog No. S72725
Change view
Click to view available options
Volume (Metric):
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S72725 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72725 Supplier LaMotte™ Supplier No. 1441

This Turbidity Standard is a calibration solution.

  • 60 mL per bottle.

Specifications

Type Turbidity Standard
For Use With (Equipment) LaMotte 2020t Turbidity Meter
Description Turbidity Standard, 1 NTU/NTRU, 60 mL
Turbidity 1 NTU/NTRU standard
Volume (English) 2 oz.
Volume (Metric) 60 mL
Packaging Type Bottle