Learn More
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU, Ricca Chemical
Turbidity Standard, Primary Standard Stock Formazin Suspension, 4000 NTU packaged in an amber glass bottle.
Specifications
Type Turbidity Standard
For Use With (Equipment) Turbidimeter
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
883016
View Documents Promotion Details
N/A 500 mL N/A
Only null left
883032
View Documents Promotion Details
N/A 1 L N/A
Only null left
R88300004C
View Documents Promotion Details
N/A 4 L N/A
Only null left
Description
Description
Turbidity Standard; Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units); Ricca Chemical; 500 mL; Amber glass
  • Turbidity Standard
  • Primary Standard Stock Formazin Suspension, 4000 NTU (Nephelometric Turbidity Units)
  • Amber glass
Specifications
Specifications
Turbidity Standard
Turbidimeter
Videos
Safety and Handling
Safety and Handling
ShelfLife : 12 Months