Learn More
Tuning Forks, Equally Tempered Scale
Catalog No. S42010M
Change view
Click to view available options
Frequency:
440 Hz
523.3 Hz
For Use With (Application):
A Scale
C Scale
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency For Use With (Application)
S42010M 523.3 Hz C Scale
S42008M 440 Hz A Scale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S42010M Supplier Customer Channels Group (Ccg) Supplier No. 7005233MU
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Eight tuning forks that resonate at natural frequencies!
The equal tempered scale divides each octave range (frequency ratio of 2:1) into 12 half steps such that every two adjacent notes have equal frequency ratios. The ratio between any two adjacent notes on this scale is the twelfth root of two or about 1.06:1. An equal tempered scale consists of two whole steps (frequency ratio 1.12:1), a half-note (frequency ratio 1.06:1), three whole steps, and a half-step. The eight tuning forks resonate at natural frequencies that coincide with the frequencies of the natural notes on the equal tempered scale. Made of aluminum alloy.
These forks are recommended by the American Association of Physics Teachers and the Acoustical Society of America for teaching purposes. All are milled from bar stock, measure 1 x 2.5cm, and are tuned to an accuracy 0.5% at 20°C. Based on A=440cps.
Compliance
Recommended by the American Association of Physics Teachers and the Acoustical Society of America for teaching purposes!
Specifications
Product Type Tuning Fork
Material Aluminum Alloy
For Use With (Application) C Scale
Frequency 523.3 Hz