Learn More
United Scientific™ Tuning Fork and Striker Set
Catalog No. S16092
Change view
Click to view available options
Product Type: Tuning Fork Set
Catalog No.
S16092 Product Type: Tuning Fork Set
1 options
Catalog No. S16092 Supplier United Scientific™ Supplier No. TFSS01
Tuning forks are available in many different frequencies.
  • Discover the science of music with the tuning fork and striker set.
  • The 256 Hz tuning fork is made of high-quality aluminum alloy and has been accurately calibrated.
  • A rubber hammer striker is included.
Specifications
Product Type Tuning Fork Set