Learn More
United Scientific™ Tuning Fork, Set of 8
Tuning forks are available in many different frequencies.
Supplier: United Scientific™ TFSET8
Description
- Convenient assortments of tuning forks.
- Made of aluminum alloy.
- The frequency and scale letter is stamped on each fork.
- Set of 8 includes a storage box.
- Set of 8 contains frequencies of 256, 288, 320, 341.3, 384, 426.6, 480, and 512 Hz.
Specifications
| Tuning Fork Set | |
| Set of 8 |