Learn More
United Scientific™ Tuning Fork Mallet, with Acrylic/Rubber Striker
Catalog No. S15715
Change view
Click to view available options
Product Type: Tuning Fork Mallet
Catalog No.
S15715 Product Type: Tuning Fork Mallet
1 options
Catalog No. S15715 Supplier United Scientific™ Supplier No. TFHAMR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For use with all types of tuning forks.
  • Tuning fork mallet has a head with one end made of hard acrylic and the other end made of rubber.
  • Head is mounted on a metal rod with an easy grip wooden handle
Specifications
Product Type Tuning Fork Mallet