Learn More
United Scientific™ Tuning Fork
Tuning forks are available in many different frequencies.
Supplier:  United Scientific™ TFK426
 View more versions of this product
Catalog No. S15857
Only null left
Explore available promotions
Description
Description
  • Made of aluminum alloy.
  • The frequency and scale letter is stamped on each fork.
  • Available in a wide variety of frequencies.
Specifications
Specifications
Tuning Fork
426.6 Hz
Product Suggestions
Product Suggestions