Learn More
United Scientific™ Tuning Fork
Catalog No. S16089
Change view
Click to view available options
Frequency:
100 Hz
1024 Hz
128 Hz
2048 Hz
256 Hz
288 Hz
320 Hz
341.3 Hz
384 Hz
4096 Hz
426.6 Hz
480 Hz
512 Hz
640 Hz
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frequency
S16089 341.3 Hz
S16033 100 Hz
S15929 128 Hz
S16030 256 Hz
S16032 288 Hz
S15830 320 Hz
S15964 384 Hz
S15857 426.6 Hz
S16064 480 Hz
S16066 512 Hz
S16069 640 Hz
S15821 1024 Hz
S15720 2048 Hz
S15963 4096 Hz
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. S16089 Supplier United Scientific™ Supplier No. TFK341
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Tuning forks are available in many different frequencies.
  • Made of aluminum alloy.
  • The frequency and scale letter is stamped on each fork.
  • Available in a wide variety of frequencies.
Specifications
Product Type Tuning Fork
Frequency 341.3 Hz