missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Tunicamycin, Calbiochem™,

$720.00 - $720.00

Chemical Identifiers

CAS 11089-65-9
Molecular Formula C39H64N4O16
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
65438050MG
View Documents
MilliporeSigma™
65438050MG
50 mg Glass Bottle
Each for $720.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

11089-65-9
C39H64N4O16

Specifications

11089-65-9
Glass Bottle
50 mg
White
C39H64N4O16
Solid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'