missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungstic(VI) acid
CAS: 7783-03-1 | H2O4W | 249.85 g/mol
$60.38 - $875.59
Chemical Identifiers
| CAS | 7783-03-1 |
|---|---|
| Molecular Formula | H2O4W |
| Molecular Weight (g/mol) | 249.85 |
| MDL Number | MFCD00011336 |
| InChI Key | CMPGARWFYBADJI-UHFFFAOYSA-L |
| Synonym | tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g |
| PubChem CID | 1152 |
| ChEBI | CHEBI:36272 |
| IUPAC Name | dihydroxy(dioxo)tungsten |
| SMILES | O[W](=O)(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA8211814
|
Thermo Scientific Chemicals
08211814 |
25 g |
Each for $60.38
|
|
|||||
|
AA8211822
|
Thermo Scientific Chemicals
08211822 |
100 g |
Each for $187.12
|
|
|||||
|
AA8211836
|
Thermo Scientific Chemicals
08211836 |
500 g |
Each for $875.59
|
|
|||||
Description
Applied as flavoring agent and fragrance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7783-03-1 | |
| 249.85 | |
| CMPGARWFYBADJI-UHFFFAOYSA-L | |
| 1152 | |
| dihydroxy(dioxo)tungsten |
| H2O4W | |
| MFCD00011336 | |
| tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g | |
| CHEBI:36272 | |
| O[W](=O)(=O)O |
Specifications
| 1473°C | |
| 7783-03-1 | |
| 25 g | |
| MFCD00011336 | |
| Insoluble in water and acid. Soluble in alkaline solution. | |
| O[W](=O)(=O)O | |
| 249.85 | |
| CHEBI:36272 | |
| 2.24 | |
| Tungstic(VI) acid |
| 100°C (decomposition) | |
| Powder | |
| H2O4W | |
| tungstic acid, tungstic vi acid, dihydroxy dioxo tungsten, dihydrogen wolframate, wolframsaure, tunstic acid, dihydroxidodioxidotungsten, tungstic acid h2wo4, unii-j4d6k0rx2g | |
| CMPGARWFYBADJI-UHFFFAOYSA-L | |
| dihydroxy(dioxo)tungsten | |
| 1152 | |
| 249.86 | |
| 5.5 |
Safety and Handling
EINECSNumber : 231-975-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only