missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungstic acid, 99+%
CAS: 7783-03-1 | H2O4W | 249.86 g/mol
$147.34 - $147.34
Chemical Identifiers
| CAS | 7783-03-1 |
|---|---|
| Molecular Formula | H2O4W |
| Molecular Weight (g/mol) | 249.86 |
| MDL Number | MFCD00011336 |
| InChI Key | CMPGARWFYBADJI-UHFFFAOYSA-L |
| IUPAC Name | dioxotungstendiol |
| SMILES | O[W](O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC207611000
|
Thermo Scientific Chemicals
207611000 |
100 g | Plastic bottle |
Each for $147.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
| 7783-03-1 | |
| 249.86 | |
| CMPGARWFYBADJI-UHFFFAOYSA-L | |
| O[W](O)(=O)=O |
| H2O4W | |
| MFCD00011336 | |
| dioxotungstendiol |
Specifications
| Green-Yellow to Yellow | |
| Powder | |
| 99% min | |
| H2WO4 | |
| 15, 9998 | |
| CMPGARWFYBADJI-UHFFFAOYSA-L | |
| dioxotungstendiol | |
| 249.86 | |
| Plastic bottle | |
| 5.5000g/mL |
| 7783-03-1 | |
| 100 g | |
| H2O4W | |
| MFCD00011336 | |
| Solubility in water: insoluble. Other solubilities: soluble in hf and alkalies | |
| O[W](O)(=O)=O | |
| 249.86 | |
| ≥99% | |
| 5.5 | |
| Tungstic acid |
Safety and Handling
EINECSNumber : 231-975-2
RUO – Research Use Only