missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten wire, gold plated, 0.05mm (0.002in) dia, W 99.95% (metals basis), Au 99.99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Tungsten wire, gold plated |
| Name Note | 0.05mm (0.002 in.) dia. |
| Quantity | 1 m |
| Weight | ≈0.02g/m (W only), 3 to 5 wt% Au plating |
| Form | Wire |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00011461 |
| Percent Purity | 99.95% |
| Concentration or Composition (by Analyte or Components) | Au 99.99% |
RUO – Research Use Only