Learn More
Tungsten(VI) oxide, 99+%
CAS: 1314-35-8 | O3W | 231.837 g/mol
$159.81 - $520.23
Chemical Identifiers
| CAS | 1314-35-8 |
|---|---|
| MDL Number | MFCD00011466 |
| InChI Key | ZNOKGRXACCSDPY-UHFFFAOYSA-N |
| Synonym | tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa |
| PubChem CID | 14811 |
| IUPAC Name | trioxotungsten |
| SMILES | O=[W](=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC214211000
|
Thermo Scientific Chemicals
214211000 |
100 g | Plastic bottle |
Each for $159.81
|
|
||||
|
AC214215000
|
Thermo Scientific Chemicals
214215000 |
500 g | Plastic bottle |
Each for $520.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1314-35-8 | |
| ZNOKGRXACCSDPY-UHFFFAOYSA-N | |
| 14811 | |
| O=[W](=O)=O |
| MFCD00011466 | |
| tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa | |
| trioxotungsten |
Specifications
| Green-Yellow to Yellow | |
| Powder | |
| 99+% | |
| WO3 | |
| 11,591 | |
| tungsten trioxide, tungsten vi oxide, tungstic oxide, tungstic anhydride, tungsten oxide wo3, tungsten blue, tungstic acid anhydride, wolframic acid, anhydride, tungstentrioxide, acmc-20ajaa | |
| O=[W](=O)=O | |
| 231.837 | |
| 231.85 | |
| Plastic bottle |
| 1314-35-8 | |
| 100 g | |
| O3W | |
| MFCD00011466 | |
| 15, 9997 | |
| ZNOKGRXACCSDPY-UHFFFAOYSA-N | |
| trioxotungsten | |
| 14811 | |
| ≥99.0% | |
| Tungsten(VI) oxide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 215-231-4
RUO – Research Use Only