missing translation for 'onlineSavingsMsg'
Learn More

Tungsten silicide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7048818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1155109 10 g
AA1155118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1155109 Supplier Thermo Scientific Chemicals Supplier No. 1155109
Only null left

Tungsten silicide is used in microelectronics as a contact material. It is also used as a shunt over polysilicon lines to increase their conductivity and increase signal speed. Further, it acts as a barrier layer between silicon and other metals. In addition to this, it is used in microelectro mechanical systems and for oxidation-resistant coatings. It is also employed as a replacement for earlier tungsten films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12039-88-2
Molecular Formula Si2W
Molecular Weight (g/mol) 240.01
MDL Number MFCD00049704
InChI Key WQJQOUPTWCFRMM-UHFFFAOYSA-N
Synonym tungsten disilicide, tungsten-silicide, tungsten silicide wsi2, tungsten silicide, w5si3
PubChem CID 16212546
IUPAC Name bis($l^{3}-silanylidyne)tungsten
SMILES [Si]#[W]#[Si]

Specifications

Melting Point 2165°C
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 240.02
Percent Purity 99.5%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tungsten silicide
EINECSNumber 234-909-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only