missing translation for 'onlineSavingsMsg'
Learn More

Tungsten dichloride dioxide, 99%

CAS: 13520-76-8 | WO2Cl2

$360.12 - $1,188.05

Chemical Identifiers

CAS 13520-76-8
Molecular Formula WO2Cl2
MDL Number MFCD00054136
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8996209
View Documents
Thermo Scientific Chemicals
08996209
10 g N/A
Only null left
 
AA8996218
View Documents
Thermo Scientific Chemicals
08996218
50 g N/A
Only null left
 
Description

Description

Tungsten dichloride dioxide is used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13520-76-8
WO2Cl2
MFCD00054136

Specifications

Yellow
Powder
99%
MFCD00054136
286.75
Moisture sensitive
13520-76-8
10 g
WO2Cl2
Soluble in cold water. Decomposes in hot water.
Acrid
Tungsten dichloride dioxide
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 236-862-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only