missing translation for 'onlineSavingsMsg'
Learn More

Tungsten carbide, 99.5% (metals basis)

Catalog No. AA4166530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4166530 250 g
AA4166518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4166530 Supplier Thermo Scientific Chemicals Supplier No. 04166530
Only null left

CAS: 12070-13-2 | CW2 | 379.69 g/mol

Tungsten carbide is used in industrial machinery, cutting tools, abrasives, armor-piercing rounds and jewelry. It is preferred even over stainless steel because of their incredible hardness and resistance to wear and tear. It is utilized for the manufacture of mill products including grinding and milling. It also includes tipping trekking, ski poles as well as cleats. It is mainly used in the form of cemented tungsten carbide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12070-13-2
Molecular Formula CW2
Molecular Weight (g/mol) 379.69
MDL Number MFCD00801036
InChI Key FHLFYDOXSZQEPO-UHFFFAOYSA-N
Synonym tungsten carbide w2c, methyliumylidyneditungstenene
PubChem CID 102601578
IUPAC Name methyliumylidyneditungstenene
SMILES [W-]=[W]#[C+]

Specifications

Melting Point ∼2800°C
Color Black
Physical Form Powder
Quantity 250 g
Linear Formula W2C
UN Number UN3178
Solubility Information Soluble in nitric acid and hydrofluoric acid. Insoluble in water.
Formula Weight 379.71
Percent Purity 99.5%
Density 17.15 g/mL
Assay (metals basis)
Chemical Name or Material Tungsten carbide
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 235-124-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only