missing translation for 'onlineSavingsMsg'
Learn More

Tungsten boride, 99% (metals basis)

CAS: 12007-09-9 | BW | 194.65 g/mol

$236.55 - $963.86

Chemical Identifiers

CAS 12007-09-9
Molecular Formula BW
Molecular Weight (g/mol) 194.65
MDL Number MFCD00049708
InChI Key JEEHQNXCPARQJS-UHFFFAOYSA-N
Synonym tungsten boride, tungsten monoboride, tungstenioborane
PubChem CID 82793
IUPAC Name boranylidynetungsten
SMILES B#[W]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1255914
View Documents
Thermo Scientific Chemicals
01255914
25 g
Each for $236.55
Only null left
 
AA1255922
View Documents
Thermo Scientific Chemicals
01255922
100 g
Each for $963.86
Only null left
 
Description

Description

it is used as an abrasives used in thin, thick Film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12007-09-9
194.65
JEEHQNXCPARQJS-UHFFFAOYSA-N
82793
B#[W]
BW
MFCD00049708
tungsten boride, tungsten monoboride, tungstenioborane
boranylidynetungsten

Specifications

12007-09-9
25 g
MFCD00049708
It is insoluble in water.
B#[W]
194.65
194.66
Odorless
(metals basis)
Powder
BW
tungsten boride, tungsten monoboride, tungstenioborane
JEEHQNXCPARQJS-UHFFFAOYSA-N
boranylidynetungsten
82793
99%
Tungsten boride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-498-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only