Learn More
California Advanced Labeling TubeWriter 360™ Direct High-Speed Labware Printing System
Catalog No. 02500200
Encompass
Change view
Click to view available options
Product Line:
TubeWriter 360
Catalog No. Product Line
02500200 TubeWriter 360
1 options
Catalog No. 02500200 Supplier California Advanced Labeling Supplier No. TW0360A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High-speed printing on any labware
TubeWriter 360 ends manual labeling and allows mistake-free automated printing.
  • High-speed direct tube printing eliminates labels and messy handwriting
  • Prints on any labware — Microcentrifuge tubes, screw cap tubes, conical tubes, snap cap tubes, cluster tubes, cryogenic vials, histology cassettes, slides and microplates
  • Scannable barcode printing on demand (e.g., 2D Data Matrix, 39, 128, Interleaved 2 of 5)
  • Excel Interface or LIMS
  • Can print graphics, logos, and in other languages
Order Info
Installation and training is also available separately. (Contact Fisher Customer Service for details).
Specifications
Product Line TubeWriter 360
Product Type Labware Printer
For Use With (Equipment) Labware
Includes Software and one year warranty