Learn More
Chemglass Inc TLC Spotting Capillary Tubes, 6in Length, Approx 1300 Pcs/Pkg
Catalog No. CG118202
Encompass
TLC Spotting Capillary Tubes, 6in Length, Approx 1300 Pcs/Pkg. Non-calibrated tubes made from borosilicate glass for use with aqueous and organic solutions. Packed in half pound packages. Tubes are 1.0-1.1mm OD x .66-.70mm ID.
Catalog No. CG118202 Supplier Chemglass Inc Supplier No. CG118202
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
TLC Spotting Capillary Tubes, 6in Length, Approx 1300 Pcs/Pkg. Non-calibrated tubes made from borosilicate glass for use with aqueous and organic solutions. Packed in half pound packages. Tubes are 1.0-1.1mm OD x .66-.70mm ID.