missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tubes and Domed Caps, strips of 8

Catalog No. p-7184431
Change view
Click to view available options
Color:
Natural
Product Type:
8-Strip Tube
Quantity:
10 Packs of 12 Strips
250 Strips
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Quantity
AB0266 Natural 8-Strip Tube 250 Strips
AB0451 Natural 8-Strip Tube 10 Packs of 12 Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AB0266 Supplier Thermo Scientific™ Supplier No. AB0266
Only null left

Optimize PCR and qPCR with these 0.2 mL strip tubes, available as 8 tubes per strip in several colors or 12 tubes per strip.

0.2 mL Strip Tubes are compatible with standard 0.2 mL thermal cycler blocks. They are designed for both PCR and qPCR applications.

Multiple cap options are available, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL.

Highlights

  • Compatible with 0.2 mL thermal cycler blocks 8 tubes per strip 0.25 mL maximum tube capacity when capped
  • Sealable using flat and domed cap strips
  • Ultra-Clear cap options available for use in qPCR assays
Applications
  • PCR
  • qPRC
  • Real-Time PCR

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Natural
Volume (Metric) Working 0.2 mL
Volume (Metric) 0.25 mL
Quantity 250 Strips
For Use With (Equipment) Thermal Cyclers
Sterility Non-sterile
Material Polypropylene
Reaction Speed Standard
Closure Type Domed Cap
No. of Wells 96
Format 250 Strip Tubes and Caps
Product Type 8-Strip Tube
Show More Show Less