Learn More
Beckman Coulter Ultra-Clear Tube, 25x89mm
Supplier:  Beckman Coulter 344058
Encompass
This round bottom tube is designed for use in Beckman Coulter instruments and rotors. Ultra-Clear tubes are transparent, allowing for easy location of visibly banded or pelleted samples.Compatible Ultracentrifuge Rotors. Fixed Angle: Type 70 Ti, Type 50.2 Ti. Swinging Bucket: SW 32 Ti, SW 28. Compatible High Performance Rotor. Swinging Bucket: JS-24.38
Catalog No. 361042342