missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Tube Fitting, Cheminert™ 1/4-28 male UNF
Fittings and washers included. Chemically inert. Zero dead volume, small bore. Autoclavable. 500 psi (35 kg/cm3) pressure rating
$68.33 - $80.35
| Catalog Number | Mfr. No. | Description | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Description | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
|
111011782
|
MilliporeSigma™ Supelco™
58712 |
For Tubing 1/8 in. O.D. | 3 mm |
Each for $80.35
|
|
||||
|
111011869
|
MilliporeSigma™ Supelco™
58705 |
For Tubing 1/16 in. O.D. | 1.5 mm |
Each for $68.33
|
|
||||
Specifications
| Mixed | |
| 12/Pk. | |
| Stainless Steel |
Cheminert is a registered trademark of Valco Instruments Co. Inc.