Learn More
Eisco™ Tube Carrier
Catalog No. S24474
Change view
Click to view available options
Quantity:
Accommodates up to 15 tubes
Catalog No. Quantity
S24474 Accommodates up to 15 tubes
1 options
Catalog No. S24474 Supplier Eisco™ Supplier No. BI0248
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For attachment to clinostat and experiments in geotropism.
  • With germinating seedlings
  • Accommodates up to 15 tubes
Specifications
Product Type Tube Carrier
For Use With (Application) Experiments in geotropism
Includes Germinating seedlings
Quantity Accommodates up to 15 tubes
Diameter (Metric) 180 mm
For Use With (Equipment) Attachment to clinostat