Learn More
Eppendorf™ Tube Adapters for Eppendorf™ Square Buckets
Fit in square bucket 05-400-309 to accept round-bottom tubes from 5 to 25mL
$418.81 - $454.00
Specifications
| For Use With (Equipment) | A-4-38 swing bucket rotor (rectangular bucket) for 5702/5702R/5702RH centrifuge |
|---|---|
| Quantity | Set of 2 |
Description
Order Info
Adapters are sold in packs of two. Be sure to order the proper number to fully equip your rotor.
Specifications
| A-4-38 swing bucket rotor (rectangular bucket) for 5702/5702R/5702RH centrifuge | |
| Set of 2 |