Learn More
Eppendorf™ Tube Adapters for Eppendorf™ Model 5702 Round Buckets
Fit in round bucket 05-400-320 to accept tubes from 1.1 to 85mL
Supplier: Eppendorf™ 022639200
Description
Adapters are sold in packs of two. Be sure to order proper number to fully equip your rotor.
Chemical Identifiers
| A-4-38 swing bucket rotor (round bucket) for 5702/5702R/5702RH centrifuge |
Specifications
| A-4-38 swing bucket rotor (round bucket) for 5702/5702R/5702RH centrifuge | |
| 15 mL Conical Tubes | |
| 2 |