Learn More
Labcon North America 50mL SuperClear Centrifuge Tubes, in Bulk DFS Item
Supplier:  Labcon North America 31918700089
Tube, Conical centrifuge; Labcon; SuperClear; 50mL; With flat screw caps; Caps include integral molded elastomeric seal; Patented anti-smear writing surface; 3 color printed graduations and reverse reading scale; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Polypropylene USP Class VI resin; Centrifuging to 20,000 x G; Freezable -80C; Autoclavable; Capacity: 50mL; Teal color caps; 500 tubes per bag/500 caps in separate bag/500 tubes and caps per case; 29mm W. 115mm H; Made in USA
Catalog No. 50-101-569
Only null left
Explore available promotions