Learn More
Labcon North America SuperClear 12x75mm Culture Tubes with Dual Position Caps, Polystyrene, 125 per Bag, Sterile
Supplier:  Labcon North America 33283550009
Encompass
Tube, Culture; Labcon; SuperClear; 5mL; Sterile; 12x75mm culture tubes with attached dual position polyethylene caps; For cell culture and general laboratory use; With extra thick walls to prevent breakage; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polystyrene USP Classs VI resin; Centrifuging to 1,500 x G; Not Freezable; Not Autoclavable; Capacity: 5mL; 125 tubes per bag/8 bags per case; 12mm W. 75mm H; Made in USA
Catalog No. 50101520
Only null left