missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab TTF-1 Thyroid IHC Control Slides
Description
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners
- Each package includes specific staining protocol
- 15 per package
- Tissue: Thyroid Special Stain or Antibody TTF-1
Specifications
Specifications
| Product Type | Control Slide |
| For Use With (Application) | Histology Laboratories |